4-tert-Butyl-2,6-dimethylaniline ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(tert-Butyl)-2,6-dimethylaniline

081190-1G 081190-10G
102834-294EA 143.08 USD
102834-294 102834-296
4-tert-Butyl-2,6-dimethylaniline ≥97%
4-tert-Butyl-2,6-dimethylaniline

Hazard: Toxic/Stored under Argon/Air Sensitive

Formula: C₁₂H₁₉N
Boiling Pt: 96°C/1mm
MDL Number: MFCD00052181
CAS Number: 42014-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR