1-(4-Aminophenyl)piperazine ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Piperazin-1-ylaniline

081179-1G 081179-10G
102834-268EA 226.15 USD
102834-268 102834-270
1-(4-Aminophenyl)piperazine ≥97%
1-(4-Aminophenyl)piperazine

Hazard: Irritant

Formula: C₁₀H₁₅N₃
MDL Number: MFCD01365906
CAS Number: 67455-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR