1,3-Bis(4-aminophenoxy)propane ≥97%

Supplier: Matrix Scientific

Synonyms: 4,4'-(Propane-1,3-diylbis(oxy))dianiline, 4,4'-(1,3-Propanediyl)dioxydianiline

081177-1G 081177-10G
102834-262EA 340 USD
102834-262 102834-264
1,3-Bis(4-aminophenoxy)propane ≥97%
1,3-Bis(4-aminophenoxy)propane

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₂
MW: 258.3200 g/mol
MDL Number: MFCD00025331
CAS Number: 52980-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR