4-[1-(4-Aminophenyl)-1-methylethyl]aniline ≥97%

Supplier: Matrix Scientific

Synonyms: 4,4'-(Propane-2,2-diyl)dianiline

081169-250MG 081169-1G
102834-240EA 231.46 USD
102834-240 102834-242
4-[1-(4-Aminophenyl)-1-methylethyl]aniline ≥97%
4-[1-(4-Aminophenyl)-1-methylethyl]aniline

Hazard: Irritant

Formula: C₁₅H₁₈N₂
MW: 226.32 g/mol
MDL Number: MFCD00438683
CAS Number: 2479-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR