2-Bromo-4,6-dinitroaniline ≥97%

Supplier: Matrix Scientific

081167-10G 081167-50G
102834-234EA 64.62 USD
102834-234 102834-236
2-Bromo-4,6-dinitroaniline ≥97%
2-Bromo-4,6-dinitroaniline

Hazard: Irritant

Formula: C₆H₄BrN₃O₄
MW: 262.02 g/mol
MDL Number: MFCD00007146
CAS Number: 1817-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR