4,6-Dichloro-o-toluidine ≥97%

Supplier: Matrix Scientific

Synonyms: 2,4-Dichloro-6-methylaniline

081161-1G 081161-10G
102834-216EA 72.31 USD
102834-216 102834-218
4,6-Dichloro-o-toluidine ≥97%
4,6-Dichloro-o-toluidine

Hazard: Irritant

Formula: C₇H₇Cl₂N
MW: 176.05 g/mol
MDL Number: MFCD00044074
CAS Number: 30273-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR