4-Chloro-3-nitroaniline ≥97%

Supplier: Matrix Scientific

081157-1G 081157-10G 081157-50G
102834-200EA 130.77 USD
102834-200 102834-202 102834-204
4-Chloro-3-nitroaniline ≥97%
4-Chloro-3-nitroaniline

Hazard: Irritant

Formula: C₆H₅ClN₂O₂
MDL Number: MFCD00007844
CAS Number: 635-22-3
UN: 2237
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR