3-Nitro-o-phenylenediamine ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Nitrobenzene-1,2-diamine, 3-Nitro-1,2-phenylenediamine

081152-1G 081152-5G
102834-188EA 78.46 USD
102834-188 102834-190
3-Nitro-o-phenylenediamine ≥97%
3-Nitro-o-phenylenediamine

Hazard: Irritant

Formula: C₆H₇N₃O₂
MDL Number: MFCD00007722
CAS Number: 3694-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR