4,6-Dibromo-2,3-dichloroaniline ≥97%

Supplier: Matrix Scientific

081150-1G 081150-10G
102834-180EA 101.54 USD
102834-180 102834-182
4,6-Dibromo-2,3-dichloroaniline ≥97%
4,6-Dibromo-2,3-dichloroaniline

Hazard: Irritant

Formula: C₆H₃Br₂Cl₂N
MW: 319.81 g/mol
MDL Number: MFCD00052815
CAS Number: 113571-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR