o-Anisidinium chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methoxyaniline hydrochloride

081148-1G 081148-10G
102834-174EA 72.31 USD
102834-174 102834-176
o-Anisidinium chloride ≥97%
o-Anisidinium chloride

Hazard: Irritant

Formula: C₇H₁₀ClNO
MW: 159.6134 g/mol
MDL Number: MFCD00054334
CAS Number: 134-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR