4-Methoxy-N-methylaniline ≥97%
Supplier: Matrix Scientific
Synonyms:
N-Methyl-p-anisidine
081147-1G
081147-10G
102834-170EA
101.54
USD
102834-170
102834-172
4-Methoxy-N-methylaniline ≥97%
4-Methoxy-N-methylaniline
Hazard: Irritant
Formula:
C₈H₁₁NO MW: 137.18 g/mol |
MDL Number:
MFCD00008399 CAS Number: 5961-59-1 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...