N-(4-Aminophenyl)-N-ethyl-N-isopropylamine hydrochloride ≥97%

Supplier: Matrix Scientific

081144-1G 081144-10G
102834-162EA 127.69 USD
102834-162 102834-164
N-(4-Aminophenyl)-N-ethyl-N-isopropylamine hydrochloride ≥97%
N-(4-Aminophenyl)-N-ethyl-N-isopropylamine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₉ClN₂
MDL Number: MFCD00211071
CAS Number: 91215-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR