Ethyl {[4-(trifluoromethoxy)phenyl]amino}acetate ≥97%

Supplier: Matrix Scientific

081143-1G 081143-10G
102834-158EA 241.54 USD
102834-158 102834-160
Ethyl {[4-(trifluoromethoxy)phenyl]amino}acetate ≥97%
Ethyl {[4-(trifluoromethoxy)phenyl]amino}acetate

Hazard: Irritant

Formula: C₁₁H₁₂F₃NO₃
MDL Number: MFCD00176763
CAS Number: 306935-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR