4-Amino-3-chlorophenol hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Amino-3-chlorophenol Hydrochloride, 2-Chloro-4-hydroxyaniline hydrochloride

081141-1G 081141-10G 081141-50G
102834-150EA 72.31 USD
102834-150 102834-152 102834-154
4-Amino-3-chlorophenol hydrochloride ≥97%
4-Amino-3-chlorophenol hydrochloride

Hazard: Irritant

Formula: C₆H₇Cl₂NO
MW: 180.04 g/mol
MDL Number: MFCD00143110
CAS Number: 52671-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR