2-Nitro-4,5-xylidine ≥97%

Supplier: Matrix Scientific

081123-1G 081123-10G 081123-50G
102834-110EA 56.92 USD
102834-110 102834-112 102834-114
2-Nitro-4,5-xylidine ≥97%
2-Nitro-4,5-xylidine

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD00007811
CAS Number: 6972-71-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR