4-Iodo-2,6-dimethylaniline ≥97%

Supplier: Matrix Scientific

081120-5G 081120-25G
102834-100EA 230.77 USD
102834-100 102834-102
4-Iodo-2,6-dimethylaniline ≥97%
4-Iodo-2,6-dimethylaniline

Hazard: Irritant/Store in Freezer

Formula: C₈H₁₀IN
MW: 247.08 g/mol
MDL Number: MFCD08061611
CAS Number: 4102-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR