2-Fluoro-N-methylaniline ≥97%

Supplier: Matrix Scientific

Synonyms: N-Methyl-2-fluoroaniline

081114-1G 081114-10G
102834-086EA 86.15 USD
102834-086 102834-088
2-Fluoro-N-methylaniline ≥97%
2-Fluoro-N-methylaniline

Hazard: Irritant

Formula: C₇H₈FN
MW: 125.15 g/mol
MDL Number: MFCD00028030
CAS Number: 1978-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR