2-Amino-p-cresol ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Amino-4-hydroxytoluene, 1-Amino-2-hydroxy-5-methylbenzene, 2-Amino-4-methylphenol, 2-Hydroxy-5-methylaniline

081111-10G 081111-50G
102834-078EA 44.62 USD
102834-078 102834-080
2-Amino-p-cresol ≥97%
2-Amino-p-cresol

Hazard: Irritant

Formula: C₇H₉NO
MW: 123.15 g/mol
MDL Number: MFCD00007699
CAS Number: 95-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR