5-Fluoro-o-anisidine ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Fluoro-2-methoxyaniline, 5-Fluoro-2-anisidine, 2-Amino-4-fluoroanisole

081107-5G 081107-25G
102834-068EA 96.92 USD
102834-068 102834-070
5-Fluoro-o-anisidine ≥97%
5-Fluoro-o-anisidine

Hazard: Toxic

Formula: C₇H₈FNO
MW: 141.15 g/mol
MDL Number: MFCD00077537
CAS Number: 1978-39-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR