2,3-Diaminotoluene ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Methylbenzene-1,2-diamine, Toluene-2,3-diamine, 1,2-Diamino-3-methylbenzene

081105-1G 081105-10G
102834-058EA 101.54 USD
102834-058 102834-060
2,3-Diaminotoluene ≥97%
2,3-Diaminotoluene

Hazard: Irritant

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD00011589
CAS Number: 2687-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR