4-Decylaniline hydrochloride 97
Supplier: Matrix Scientific
081103-1G
081103-10G
102834-054EA
143.08
USD
102834-054
102834-056
4-Decylaniline hydrochloride 97
4-Decylaniline hydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₈ClN MW: 269.86 g/mol |
MDL Number:
MFCD00155303 CAS Number: 175136-55-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...