2,5-Xylidine ≥98%
Supplier: Matrix Scientific
Synonyms:
2-Amino-p-xylene, 2,5-Dimethylaniline
081099-10G
081099-50G
102834-042EA
46.15
USD
102834-042
102834-044
2,5-Xylidine ≥98%
2,5-Xylidine
Hazard: Irritant
Formula:
C₈H₁₁N MW: 121.18 g/mol |
MDL Number:
MFCD00007743 CAS Number: 95-78-3 UN: 1711 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...