Bromoacetyl chloride ≥97%

Supplier: Matrix Scientific

081098-10G 081098-50G
102834-038EA 90.77 USD
102834-038 102834-040
Bromoacetyl chloride ≥97%
Bromoacetyl chloride

Hazard: Corrosive

Formula: BrCH₂COOCl
MW: 157.39 g/mol
Boiling Pt: 125…127 °C
MDL Number: MFCD00000724
CAS Number: 22118-09-8
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR