2-Chloro-4'-phenylacetophenone ≥97%
Supplier: Matrix Scientific
Synonyms:
1-([1,1-Biphenyl]-4-yl)-2-chloroethanone
081096-1G
081096-10G
102834-032EA
184.62
USD
102834-032
102834-034
2-Chloro-4'-phenylacetophenone ≥97%
2-Chloro-4'-phenylacetophenone
Hazard: Irritant
Formula:
C₁₄H₁₁ClO MW: 230.6895 g/mol |
MDL Number:
MFCD00058937 CAS Number: 635-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...