2-Chloro-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethanone ≥95%

Supplier: Matrix Scientific

081095-1G 081095-10G
102834-028EA 101.54 USD
102834-028 102834-030
2-Chloro-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethanone ≥95%
2-Chloro-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethanone

Hazard: Irritant

Formula: C₁₀H₉ClO₃
MW: 212.63 g/mol
MDL Number: MFCD00099407
CAS Number: 93439-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR