2-Bromo-1-[3-(2,4-dichlorophenyl)isoxazol-5-yl]ethanone ≥97%
Supplier: Matrix Scientific
081090-1G
081090-10G
102834-016EA
241.54
USD
102834-016
102834-018
2-Bromo-1-[3-(2,4-dichlorophenyl)isoxazol-5-yl]ethanone ≥97%
2-Bromo-1-[3-(2,4-dichlorophenyl)isoxazol-5-yl]ethanone
Hazard: Irritant
Formula:
C₁₁H₆BrCl₂NO₂ MW: 334.99 g/mol |
MDL Number:
MFCD00173932 CAS Number: 175334-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...