2-Bromo-1-(pyridin-4-yl)ethanone hydrobromide ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Bromoacetyl)pyridine hydrobromide

081087-1G 081087-10G
102834-008EA 69.23 USD
102834-008 102834-010
2-Bromo-1-(pyridin-4-yl)ethanone hydrobromide ≥95%
2-Bromo-1-(pyridin-4-yl)ethanone hydrobromide

Hazard: Irritant

Formula: C₇H₇Br₂NO
MW: 280.95
Melting Pt: ∼200 °C
MDL Number: MFCD02681893
CAS Number: 5349-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR