ω-Bromo-3,4-dichloroacetophenone ≥97%

Supplier: Matrix Scientific

081080-1G 081080-10G
102833-988EA 72.31 USD
102833-988 102833-990
ω-Bromo-3,4-dichloroacetophenone ≥97%
ω-Bromo-3,4-dichloroacetophenone

Hazard: Irritant

Formula: C₈H₅BrCl₂O
Melting Pt: 58°C
MDL Number: MFCD00051581
CAS Number: 2632-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR