2-Bromo-1-(4-chloro-3-methylphenyl)ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-3-methylphenacyl bromide

081077-1G 081077-10G
102833-978EA 213.85 USD
102833-978 102833-980
2-Bromo-1-(4-chloro-3-methylphenyl)ethanone ≥95%
2-Bromo-1-(4-chloro-3-methylphenyl)ethanone

Hazard: Irritant

Formula: C₉H₈BrClO
MW: 247.53 g/mol
MDL Number: MFCD00051586
CAS Number: 205178-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR