2-Hydroxy-5-aminopyridine hydrochloride ≥98%
Supplier: Matrix Scientific
081056-1G
081056-5G
102672-564EA
61.54
USD
102672-564
102672-566
2-Hydroxy-5-aminopyridine hydrochloride ≥98%
2-Hydroxy-5-aminopyridine hydrochloride
Hazard: Irritant
Formula:
C₅H₇ClN₂O MW: 146.5800 g/mol |
MDL Number:
MFCD04112744 CAS Number: 117865-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...