3,5-Dibromo-2-methylpyridine ≥98%

Supplier: Matrix Scientific

081057-1G 081057-5G
102672-568EA 186.15 USD
102672-568 102672-570
3,5-Dibromo-2-methylpyridine ≥98%
3,5-Dibromo-2-methylpyridine

Hazard: Irritant

Formula: C6H5Br2N
MW: 250.9186 g/mol
MDL Number: MFCD00234159
CAS Number: 38749-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR