10-Bromodec-1-ene ≥97%

Supplier: Matrix Scientific

081055-5G 081055-25G
102672-560EA 73.85 USD
102672-560 102672-562
10-Bromodec-1-ene ≥97%
10-Bromodec-1-ene

Hazard: Irritant

Formula: C₁₀H₁₉Br
MDL Number: MFCD00078660
CAS Number: 62871-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR