9-Bromo-1-nonene ≥97%

Supplier: Matrix Scientific

081054-5G 081054-25G
102672-556EA 63.08 USD
102672-556 102672-558
9-Bromo-1-nonene ≥97%
9-Bromo-1-nonene

Hazard: Irritant

Formula: C₉H₁₇Br
MW: 205.139 g/mol
Boiling Pt: 100-101°C/15mm
MDL Number: MFCD09037826
CAS Number: 89359-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR