3-(Acetylamino)butanoic acid

Supplier: Matrix Scientific

081047-500MG 081047-1G 081047-5G
102672-530EA 387.69 USD
102672-530 102672-532 102672-534
3-(Acetylamino)butanoic acid
3-(Acetylamino)butanoic acid

Hazard: Irritant

Formula: C₆H₁₁NO₃
MDL Number: MFCD00045919
CAS Number: 136781-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR