1-Propylpiperazin-2-one

Supplier: Matrix Scientific

081042-500MG 081042-1G 081042-5G
102672-508EA 516.92 USD
102672-508 102672-510 102672-512
1-Propylpiperazin-2-one
1-Propylpiperazin-2-one

Hazard: Irritant

Formula: C₇H₁₄N₂O
MDL Number: MFCD14707896
CAS Number: 65464-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR