2-Bromo-5-isopropyl-1,3,4-thiadiazole

Supplier: Matrix Scientific

081034-500MG 081034-1G
102672-478EA 516.92 USD
102672-478 102672-480
2-Bromo-5-isopropyl-1,3,4-thiadiazole
2-Bromo-5-isopropyl-1,3,4-thiadiazole

Hazard: Irritant

Formula: C₅H₇BrN₂S
MDL Number: MFCD09965539
CAS Number: 1019111-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR