[1,2,4]Triazolo[4,3-a]pyridine-8-carboxylic acid

Supplier: Matrix Scientific

081023-500MG 081023-1G
102672-428EA 646.15 USD
102672-428 102672-430
[1,2,4]Triazolo[4,3-a]pyridine-8-carboxylic acid
[1,2,4]Triazolo[4,3-a]pyridine-8-carboxylic acid

Hazard: Irritant

Formula: C₇H₅N₃O₂
MDL Number: MFCD07377043
CAS Number: 1216218-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR