Methyl-2-amino-4-ethyl-1,3-thiazole-5-carboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-4-ethylthiazole-5-carboxylate

081020-500MG 081020-1G 081020-5G
102672-420EA 387.69 USD
102672-420 102672-422 102672-424
Methyl-2-amino-4-ethyl-1,3-thiazole-5-carboxylate
Methyl-2-amino-4-ethyl-1,3-thiazole-5-carboxylate

Hazard: Irritant

Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
MDL Number: MFCD04064731
CAS Number: 302964-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR