4,4,4-Trifluorobutanoyl chloride

Supplier: Matrix Scientific

Synonyms: 4,4,4-TRifluorobutanoyl chloride

081017-500MG 081017-1G 081017-5G
102672-402EA 223.08 USD
102672-402 102672-404 102672-406
4,4,4-Trifluorobutanoyl chloride
4,4,4-Trifluorobutanoyl chloride

Hazard: Irritant

Formula: C₄H₄ClF₃O
MW: 160.52 g/mol
MDL Number: MFCD00092810
CAS Number: 406-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR