4-(Aminomethyl)-N,N-dimethylbenzamide hydrochloride

Supplier: Matrix Scientific

081007-500MG 081007-1G 081007-5G
102672-346EA 516.92 USD
102672-346 102672-348 102672-350
4-(Aminomethyl)-N,N-dimethylbenzamide hydrochloride
4-(Aminomethyl)-N,N-dimethylbenzamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O
MW: 214.7 g/mol
MDL Number: MFCD09836259
CAS Number: 104566-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR