1-(4-Methyl-1,3-thiazol-5-yl)ethanone

Supplier: Matrix Scientific

Synonyms: 1-(4-Methylthiazol-5-yl)ethanone

081006-500MG 081006-1G
102672-342EA 516.92 USD
102672-342 102672-344
1-(4-Methyl-1,3-thiazol-5-yl)ethanone
1-(4-Methyl-1,3-thiazol-5-yl)ethanone

Hazard: Irritant

Formula: C₆H₇NOS
MW: 141.19 g/mol
MDL Number: MFCD09029046
CAS Number: 38205-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR