3-(Chloromethyl)-5-isopropyl-1,2,4-oxadiazole

Supplier: Matrix Scientific

080998-500MG 080998-1G 080998-5G
102672-302EA 387.69 USD
102672-302 102672-304 102672-306
3-(Chloromethyl)-5-isopropyl-1,2,4-oxadiazole
3-(Chloromethyl)-5-isopropyl-1,2,4-oxadiazole

Hazard: Irritant

Formula: C₆H₉ClN₂O
MW: 160.6 g/mol
MDL Number: MFCD09864313
CAS Number: 189130-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR