3-(1H-Imidazol-2-yl)pyridine

Supplier: Matrix Scientific

080997-500MG 080997-1G 080997-5G
102672-296EA 230.77 USD
102672-296 102672-298 102672-300
3-(1H-Imidazol-2-yl)pyridine
3-(1H-Imidazol-2-yl)pyridine

Hazard: Irritant

Formula: C₈H₇N₃
MDL Number: MFCD08458369
CAS Number: 13570-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR