4-(1H-Imidazol-2-yl)pyridine
Supplier: Matrix Scientific
080996-500MG
080996-1G
080996-5G
102672-290EA
230.77
USD
102672-290
102672-292
102672-294
4-(1H-Imidazol-2-yl)pyridine
4-(1H-Imidazol-2-yl)pyridine
Hazard: Irritant
Formula:
C₈H₇N₃ |
MDL Number:
MFCD08668556 CAS Number: 21202-42-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...