3-(Bromomethyl)-5-methyl-1,2,4-oxadiazole

Supplier: Matrix Scientific

080990-500MG 080990-1G 080990-5G
102672-258EA 646.15 USD
102672-258 102672-260 102672-262
3-(Bromomethyl)-5-methyl-1,2,4-oxadiazole
3-(Bromomethyl)-5-methyl-1,2,4-oxadiazole

Hazard: Irritant

Formula: C₄H₅BrN₂O
MW: 177.01 g/mol
MDL Number: MFCD16250353
CAS Number: 959406-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR