Methyl-4-methoxybutanoate

Supplier: Matrix Scientific

Synonyms: Methyl 4-methoxybutanoate

080986-500MG 080986-1G 080986-5G
102672-238EA 387.69 USD
102672-238 102672-240 102672-242
Methyl-4-methoxybutanoate
Methyl-4-methoxybutanoate

Hazard: Irritant

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
MDL Number: MFCD00274316
CAS Number: 29006-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR