5-Chloro-1,3-dimethyl-1H-1,2,4-triazole

Supplier: Matrix Scientific

080982-500MG 080982-1G
102672-220EA 646.15 USD
102672-220 102672-222
5-Chloro-1,3-dimethyl-1H-1,2,4-triazole
5-Chloro-1,3-dimethyl-1H-1,2,4-triazole

Hazard: Irritant

Formula: C₄H₆ClN₃
MDL Number: MFCD22422030
CAS Number: 56616-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR