3-Chloro-1,5-dimethyl-1H-1,2,4-triazole

Supplier: Matrix Scientific

080981-500MG 080981-1G 080981-5G
102672-214EA 646.15 USD
102672-214 102672-216 102672-218
3-Chloro-1,5-dimethyl-1H-1,2,4-triazole
3-Chloro-1,5-dimethyl-1H-1,2,4-triazole

Hazard: Irritant

Formula: C₄H₆ClN₃
MDL Number: MFCD22422029
CAS Number: 56616-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR