Ethyl-2-(2-methyl-1,3-dioxolan-2-yl)acetate

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(2-methyl-1,3-dioxolan-2-yl)acetate, 2-(2-Methyl-1,3-dioxolan-2-yl)acetic Acid Ethyl Ester

080978-500MG 080978-1G 080978-5G
102672-196EA 306.15 USD
102672-196 102672-198 102672-200
Ethyl-2-(2-methyl-1,3-dioxolan-2-yl)acetate
Ethyl-2-(2-methyl-1,3-dioxolan-2-yl)acetate

Hazard: Irritant

Formula: C₈H₁₄O₄
MW: 174.20 g/mol
Boiling Pt: 107 °C (11 Torr)
Density: 1.0409/20°C
MDL Number: MFCD00152488
CAS Number: 6413-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR