2-{[4-(1-Methyl-1-phenylethyl)phenoxy]methyl}oxirane

Supplier: Matrix Scientific

080977-500MG 080977-1G 080977-5G
102672-190EA 161.54 USD
102672-190 102672-192 102672-194
2-{[4-(1-Methyl-1-phenylethyl)phenoxy]methyl}oxirane
2-{[4-(1-Methyl-1-phenylethyl)phenoxy]methyl}oxirane

Hazard: Irritant

Formula: C₁₈H₂₀O₂
MDL Number: MFCD06660497
CAS Number: 61578-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR